Learn More
Cole-Parmer Techne MCS-104S Biological Stirrer, 4-Position, Small; 110/230 V
Catalog No. 501960480 Shop All Cole-Parmer Products
Encompass_Preferred
The soft-start/stop feature provides slow acceleration and deceleration of the stirrer, avoiding excessive turbulence in the culture media and eliminating cell damage. Use the interval stirring mode during the attachment phase to further reduce agitation of the media or when culturing particularly fragile cells. The stirrers create virtually no heat making them suitable for use in both incubators and cold rooms. The culture vessels incorporate a unique base design which, together with the bulb-ended stirrer, ensures that the cells are lifted into suspension at the lowest possible speeds. This gentle stirring action promotes high cell yields by preventing cell shearing. Order Techne cell cultrure vessels separately.
Catalog No. 50-196-0480 Supplier Cole-Parmer Supplier No. 2183703
The soft-start/stop feature provides slow acceleration and deceleration of the stirrer, avoiding excessive turbulence in the culture media and eliminating cell damage. Use the interval stirring mode during the attachment phase to further reduce agitation of the media or when culturing particularly fragile cells. The stirrers create virtually no heat making them suitable for use in both incubators and cold rooms. The culture vessels incorporate a unique base design which, together with the bulb-ended stirrer, ensures that the cells are lifted into suspension at the lowest possible speeds. This gentle stirring action promotes high cell yields by preventing cell shearing. Order Techne cell cultrure vessels separately.