Learn More
Chemglass Life Sciences Stirrer Blade, PTFE, Banana Shape, Perforated, 19mm, Heavy Duty, for 20000mL Flasks, 55/50 minimum joint clearance
SDP
Catalog No. CG209016
Encompass_Preferred
Stirrer Blade, PTFE, Banana Shape, Perforated, 19mm, Heavy Duty, for 22000mL Flasks, 55/50 minimum joint clearance. Blade is shaped to resemble the contour of a round bottom flask. Blades are perforated. Made of 3/16" PTFE to fit 19mm OD stirrer shafts. Minimum joint size listed is the size required to allow passage of the blade.
Catalog No. CG209016 Supplier Chemglass Life Sciences Supplier No. CG209016
Stirrer Blade, PTFE, Banana Shape, Perforated, 19mm, Heavy Duty, for 22000mL Flasks, 55/50 minimum joint clearance. Blade is shaped to resemble the contour of a round bottom flask. Blades are perforated. Made of 3/16" PTFE to fit 19mm OD stirrer shafts. Minimum joint size listed is the size required to allow passage of the blade.