Learn More
Chemglass Life Sciences Stirrer Blade, PTFE, Banana Shape, Perforated, 10mm, for 12000mL Flasks, 45/50 minimum joint clearance
SDP
Catalog No. CG208115
Encompass_Preferred
Stirrer Blade, PTFE, Banana Shape, Perforated, 10mm, for 12000mL Flasks, 45/50 minimum joint clearance. Blade is shaped to resemble the contour of a round bottom flask. Blades are perforated. Made of 1/8" PTFE to fit 10mm OD stirrer shafts. Minimum joint size listed is the size required to allow passage of the blade.
Catalog No. CG208115 Supplier Chemglass Life Sciences Supplier No. CG208115
Stirrer Blade, PTFE, Banana Shape, Perforated, 10mm, for 12000mL Flasks, 45/50 minimum joint clearance. Blade is shaped to resemble the contour of a round bottom flask. Blades are perforated. Made of 1/8" PTFE to fit 10mm OD stirrer shafts. Minimum joint size listed is the size required to allow passage of the blade.