Learn More
Chemglass Life Sciences Stirrer Blade, PTFE, Banana Shaped, Solid, 19mm, Heavy Duty, for 12000mL Flasks, 45/50 minimum joint clearance
SDP
Catalog No. CG209005
Encompass_Preferred
Stirrer Blade, PTFE, Banana Shaped, Solid, 19mm, Heavy Duty, for 12000mL Flasks, 45/50 minimum joint clearance. Blade is shaped to resemble the contour of a round bottom flask. Blades are solid. Made of 3/16" PTFE to fit 19mm OD stirrer shafts. Minimum joint size listed is the size required to allow passage of the blade.
Catalog No. CG209005 Supplier Chemglass Life Sciences Supplier No. CG209005
Stirrer Blade, PTFE, Banana Shaped, Solid, 19mm, Heavy Duty, for 12000mL Flasks, 45/50 minimum joint clearance. Blade is shaped to resemble the contour of a round bottom flask. Blades are solid. Made of 3/16" PTFE to fit 19mm OD stirrer shafts. Minimum joint size listed is the size required to allow passage of the blade.