Learn More
Chemglass Life Sciences Stirrer Blade, PTFE, Banana Shape, Solid, 10mm, for 20000mL Flasks, 55/50 minimum joint clearance
SDP
Catalog No. 50872512
Encompass_Preferred
Stirrer Blade, PTFE, Banana Shape, Solid, 10mm, for 22000mL Flasks, 55/50 minimum joint clearance. Blade is shaped to resemble the contour of a round bottom flask. Blades are solid. Made of 1/8" PTFE to fit 10mm OD stirrer shafts. Minimum joint size listed is the size required to allow passage of the blade.
Catalog No. 50-872-512 Supplier Chemglass Life Sciences Supplier No. CG208107
Stirrer Blade, PTFE, Banana Shape, Solid, 10mm, for 22000mL Flasks, 55/50 minimum joint clearance. Blade is shaped to resemble the contour of a round bottom flask. Blades are solid. Made of 1/8" PTFE to fit 10mm OD stirrer shafts. Minimum joint size listed is the size required to allow passage of the blade.