Learn More
Chemglass Life Sciences Stirrer Blade, PTFE, 10mm, for 250mL to 500mL Flasks, 19mm Width x 56mm Length, 24/40 minimum joint clearance
SDP
Catalog No. CG208002
Encompass_Preferred
Stirrer Blade, PTFE, 10mm, for 250mL to 500mL Flasks, 19mm Width x 60mm Length, 24/40 minimum joint clearance. Stirrer blade made of 1/8" thick solid PTFE for use with both Chem-Stir and button style stirring shafts having a 10mm OD.
Catalog No. CG208002 Supplier Chemglass Life Sciences Supplier No. CG208002
Stirrer Blade, PTFE, 10mm, for 250mL to 500mL Flasks, 19mm Width x 60mm Length, 24/40 minimum joint clearance. Stirrer blade made of 1/8" thick solid PTFE for use with both Chem-Stir and button style stirring shafts having a 10mm OD.