Learn More
Chemglass Life Sciences Stirrer Blade, PTFE, 10mm, for 20000mL Flasks, 24mm Width x 160mm Length, 29/42 minimum joint clearance
SDP
Catalog No. CG208006
Encompass_Preferred
Stirrer Blade, PTFE, 10mm, for 22000mL Flasks, 24mm Width x 160mm Length, 29/42 minimum joint clearance. Stirrer blade made of 1/8" thick solid PTFE for use with both Chem-Stir and button style stirring shafts having a 10mm OD.
Catalog No. CG208006 Supplier Chemglass Life Sciences Supplier No. CG208006
Stirrer Blade, PTFE, 10mm, for 22000mL Flasks, 24mm Width x 160mm Length, 29/42 minimum joint clearance. Stirrer blade made of 1/8" thick solid PTFE for use with both Chem-Stir and button style stirring shafts having a 10mm OD.