Learn More
Chemglass Life Sciences Stirrer Blade, PTFE, 19mm, Heavy Duty, for 50000mL Flasks, 44mm Width x 175mm Length, 55/50 minimum joint clearance
SDP
Catalog No. CG208906
Encompass_Preferred
Stirrer Blade, PTFE, 19mm, Heavy Duty, for 50000mL Flasks, 44mm Width x 175mm Length, 55/50 minimum joint clearance. Stirrer blade made of 3/16" thick solid PTFE for use with both Chem-Stir™ and button style stirring shafts having a 19mm OD.
Catalog No. CG208906 Supplier Chemglass Life Sciences Supplier No. CG208906
Stirrer Blade, PTFE, 19mm, Heavy Duty, for 50000mL Flasks, 44mm Width x 175mm Length, 55/50 minimum joint clearance. Stirrer blade made of 3/16" thick solid PTFE for use with both Chem-Stir™ and button style stirring shafts having a 19mm OD.