Learn More
Chemglass Life Sciences Stirrer Assembly, Macro, Chem-Stir, 19mm x 75mm Blade Fits 500mL-5L Flask, 29/42 Bearing, 10mm OD x 445mm Stirrer Shaft
SDP
Catalog No. CG206803
Encompass_Preferred
Stirrer Assembly, Macro, Chem-Stir, 19mm x 75mm Blade Fits 500mL-5L Flask, 29/42 Bearing, 10mm OD x 445mm Stirrer Shaft. Assembly consists of a 10mm OD Chem-Stir™ stirrer shaft, a precision bore-bearing having a standard taper inner joint and a PTFE blade. The blade is sized to fit the listed flask capacities.
Catalog No. CG206803 Supplier Chemglass Life Sciences Supplier No. CG206803
Stirrer Assembly, Macro, Chem-Stir, 19mm x 75mm Blade Fits 500mL-5L Flask, 29/42 Bearing, 10mm OD x 445mm Stirrer Shaft. Assembly consists of a 10mm OD Chem-Stir™ stirrer shaft, a precision bore-bearing having a standard taper inner joint and a PTFE blade. The blade is sized to fit the listed flask capacities.