Learn More
Chemglass Life Sciences Stirrer Assembly, Macro, Chem-Stir, 24mm x 110mm Blade Fits 2L-20L Flask, 45/50 Bearing, 10mm OD x 515mm Stirrer Shaft
SDP
Catalog No. 50873393
Encompass_Preferred
Stirrer Assembly, Macro, Chem-Stir, 24mm x 110mm Blade Fits 2L-22L Flask, 45/50 Bearing, 10mm OD x 515mm Stirrer Shaft. Assembly consists of a 10mm OD Chem-Stir stirrer shaft, a precision bore-bearing having a standard taper inner joint and a PTFE blade. The blade is sized to fit the listed flask capacities.
Catalog No. 50-873-393 Supplier Chemglass Life Sciences Supplier No. CG206805
Stirrer Assembly, Macro, Chem-Stir, 24mm x 110mm Blade Fits 2L-22L Flask, 45/50 Bearing, 10mm OD x 515mm Stirrer Shaft. Assembly consists of a 10mm OD Chem-Stir stirrer shaft, a precision bore-bearing having a standard taper inner joint and a PTFE blade. The blade is sized to fit the listed flask capacities.