Learn More
Chemglass Life Sciences Stirrer, Magnetic, Two Position, Dura-Mag, with Factory Installed Alarm, 120V
SDP
Catalog No. 50143329
Encompass
Stirrer, Magnetic, Two Position, Dura-Mag, with Factory Installed Alarm, 120V. Dura-Mag™ digital, front-end, magnetic stirrers are designed for both cell culture in spinner flasks as well as general laboratory procedures requiring the use of magnetic stirring equipment. Dura-Mag™ stirrers feature brushless, maintenance-free DC motors with drive assemblies designed for quiet operation. Stainless steel covers are stamped to show the vessel positions for accurate placement and optimum performance. Dura-Mag™ 2-position, low profile chassis measure 9.5" W x 22.5" D x 3.5” H(front to back measurement includes power cord and control knob) and are designed to fit side-by-side in most incubators. Maximum flask size is (2) 8L vessels. Optional factory installed motion alarm available. Speed range: 25-400RPM.
Catalog No. 50-143-329 Supplier Chemglass Life Sciences Supplier No. CLS4100A2
Stirrer, Magnetic, Two Position, Dura-Mag, with Factory Installed Alarm, 120V. Dura-Mag™ digital, front-end, magnetic stirrers are designed for both cell culture in spinner flasks as well as general laboratory procedures requiring the use of magnetic stirring equipment. Dura-Mag™ stirrers feature brushless, maintenance-free DC motors with drive assemblies designed for quiet operation. Stainless steel covers are stamped to show the vessel positions for accurate placement and optimum performance. Dura-Mag™ 2-position, low profile chassis measure 9.5" W x 22.5" D x 3.5” H(front to back measurement includes power cord and control knob) and are designed to fit side-by-side in most incubators. Maximum flask size is (2) 8L vessels. Optional factory installed motion alarm available. Speed range: 25-400RPM.