Learn More
Chemglass Life Sciences Stirrer, Magnetic, Five Position, Compact, Dura-Mag, 120V, with Factory Installed Alarm
SDP
Catalog No. 50143346
Encompass
Stirrer, Magnetic, Five Position, Compact, Dura-Mag, 120V, with Factory Installed Alarm. Dura-Mag digital, front-end, magnetic stirrers are designed for both cell culture in spinner flasks as well as general laboratory procedures requiring the use of magnetic stirring equipment. Dura-Mag stirrers feature brushless, maintenance-free DC motors with drive assemblies designed for quiet operation. Stainless steel covers are stamped to show the vessel positions for accurate placement and optimum performance. Optional factory installed motion alarm available. Speed range: 25-400RPM.
Catalog No. 50-143-346 Supplier Chemglass Life Sciences Supplier No. CLS4100A4
Stirrer, Magnetic, Five Position, Compact, Dura-Mag, 120V, with Factory Installed Alarm. Dura-Mag digital, front-end, magnetic stirrers are designed for both cell culture in spinner flasks as well as general laboratory procedures requiring the use of magnetic stirring equipment. Dura-Mag stirrers feature brushless, maintenance-free DC motors with drive assemblies designed for quiet operation. Stainless steel covers are stamped to show the vessel positions for accurate placement and optimum performance. Optional factory installed motion alarm available. Speed range: 25-400RPM.