Learn More
Chemglass Life Sciences Stirrer, Magnetic, Compact, Digital, Low Profile, with Timer, 100-240V, 50-60Hz
SDP
Catalog No. 501531387
Encompass_Preferred
Stirrer, Magnetic, Compact, Digital, Low Profile, with Timer, 100-240V, 50-60Hz. The MST is a compact, low profile, digital stirrer with a manually adjustable speed range of 80-1500 RPM. The bright, digital display allows for very accurate setting of the stirring speed which is adjustable in 10 rpm increments. SpeedServo technology provides for constant stirring speeds even during viscosity changes.
Catalog No. 50-153-1387 Supplier Chemglass Life Sciences Supplier No. CG2000V10
Stirrer, Magnetic, Compact, Digital, Low Profile, with Timer, 100-240V, 50-60Hz. The MST is a compact, low profile, digital stirrer with a manually adjustable speed range of 80-1500 RPM. The bright, digital display allows for very accurate setting of the stirring speed which is adjustable in 10 rpm increments. SpeedServo technology provides for constant stirring speeds even during viscosity changes.