Learn More
Cole-Parmer Mono-Mold Standard Stir Bar with Spinning Ring, 1-1/2"L x 3/8" dia
Catalog No. 50200162 Shop All Cole-Parmer Products
Encompass_Preferred
Mono-Mold magnetic stir bars are the standard all-purpose stir bar. Available with or without spinning rings; rings reduce contact area between stir bar and vessel, reducing friction and noise.
Catalog No. 50-200-162 Supplier Cole-Parmer Supplier No. 0855100
Mono-Mold magnetic stir bars are the standard all-purpose stir bar. Available with or without spinning rings; rings reduce contact area between stir bar and vessel, reducing friction and noise.