Learn More
Chemglass Life Sciences Stirrer, Magnetic, Five Position, Compact, Dura-Mag, 120V
SDP
Catalog No. 50143371
Encompass
Stirrer, Magnetic, Five Position, Compact, Dura-Mag, 120V. Dura-Mag™ digital, front-end, magnetic stirrers are designed for both cell culture in spinner flasks as well as general laboratory procedures requiring the use of magnetic stirring equipment. Dura-Mag™ stirrers feature brushless, maintenance-free DC motors with drive assemblies designed for quiet operation. Stainless steel covers are stamped to show the vessel positions for accurate placement and optimum performance. Optional factory installed motion alarm available. Speed range: 25-400RPM.
Catalog No. 50-143-371 Supplier Chemglass Life Sciences Supplier No. CLS410004
Stirrer, Magnetic, Five Position, Compact, Dura-Mag, 120V. Dura-Mag™ digital, front-end, magnetic stirrers are designed for both cell culture in spinner flasks as well as general laboratory procedures requiring the use of magnetic stirring equipment. Dura-Mag™ stirrers feature brushless, maintenance-free DC motors with drive assemblies designed for quiet operation. Stainless steel covers are stamped to show the vessel positions for accurate placement and optimum performance. Optional factory installed motion alarm available. Speed range: 25-400RPM.