Learn More
Chemglass Life Sciences Stirrer, Magnetic, Nine Position, Dura-Mag, 120V
SDP
Catalog No. 50143401
Encompass
Trouble-Free Brushless MotorIntegrated Power Supply, for 100 to 220V ServiceHeavy Duty Magnets and Drive SystemTrouble-Free Non-Slip Toothed Kevlar Belt Design with Easy ReplacementStainless Steel Stirrer Plate CoverSpeed Range from 25 to 400RPM with Clockwise ControlDimensions: 16.75" W x 19.25" D x 3.5" H(front to back depth measurement includes power cord and control knob).The Chemglass Life Sciences Nine-Position magnetic stirrer is a rugged design for 24/7 laboratory or production applications.The small footprint is ideal for CO2 incubators with maximum capacity of (9) 500mL spinner vessels.Optional factory installed motion alarm available.Please note: An Optional Flask Centering Template for Corning Disposable 500mL Spinner flasks is available.Installing the drop on template ensures the Corning Disposable Spinner flasks are centered directly over the magnet for optimal stirring.
Catalog No. 50-143-401 Supplier Chemglass Life Sciences Supplier No. CLS410009
Trouble-Free Brushless MotorIntegrated Power Supply, for 100 to 220V ServiceHeavy Duty Magnets and Drive SystemTrouble-Free Non-Slip Toothed Kevlar Belt Design with Easy ReplacementStainless Steel Stirrer Plate CoverSpeed Range from 25 to 400RPM with Clockwise ControlDimensions: 16.75" W x 19.25" D x 3.5" H(front to back depth measurement includes power cord and control knob).The Chemglass Life Sciences Nine-Position magnetic stirrer is a rugged design for 24/7 laboratory or production applications.The small footprint is ideal for CO2 incubators with maximum capacity of (9) 500mL spinner vessels.Optional factory installed motion alarm available.Please note: An Optional Flask Centering Template for Corning Disposable 500mL Spinner flasks is available.Installing the drop on template ensures the Corning Disposable Spinner flasks are centered directly over the magnet for optimal stirring.