Learn More
Hach Company Stirrer, Magnetic, Battery-Powered
Supplier:  Hach Company 2163600
Encompass
Portable, light-weight stirring ideal for field use where no electricity is available. It can be used for up to 24 hours of continuous operation, mixing up to 1 liter with a maximum stirring speed of 900 rpm using one "D" sized battery. Continuous operation time greatly extended if two "D" sized batteries are used. Dimensions: 9 ¼" x 5" x 3 ½"H. Note: For optimal performans, do not use stirring bars longer than 1 inch.
Catalog No. 50-199-5485