Learn More
Chemglass Life Sciences Stirrer, Magnetic, Compact, Analog, Low Profile, 100-240V, 50-60Hz
SDP
Catalog No. 5000980601
Encompass_Preferred
Powerful compact low profile single position analog stirrer has a manually adjustable speed range up to 1100 rpm. Technopolymer casing resists chemicals. White surface for visualization of particles and color changes. Long life operation. Reliable Results. Speed regulation up to 1100 rpm. Accepts up to 5 liter flasks. Remains cold, even after days of continuous use.
Catalog No. 50-009-80601 Supplier Chemglass Life Sciences Supplier No. CG2002V10
Powerful compact low profile single position analog stirrer has a manually adjustable speed range up to 1100 rpm. Technopolymer casing resists chemicals. White surface for visualization of particles and color changes. Long life operation. Reliable Results. Speed regulation up to 1100 rpm. Accepts up to 5 liter flasks. Remains cold, even after days of continuous use.