Learn More
Chemglass Life Sciences Support, Stirrer Stand
SDP
Catalog No. 501311197
Encompass_Preferred
Support, Stirrer Stand. Support Stand was designed to accommodate vessels up to 10" diameter, to sit close to the support rod. Support rod is ideal for mounting stirring motors. Each of the four corners rests on a rubber cushion. The powder coated steel base measures 7.875” (200mm) W x 12.5" (316mm) deep overall and has a black enamel finish. Support rod is .625” (16mm) diameter x 30” long.
Catalog No. 50-131-1197 Supplier Chemglass Life Sciences Supplier No. CG203750
Support, Stirrer Stand. Support Stand was designed to accommodate vessels up to 10" diameter, to sit close to the support rod. Support rod is ideal for mounting stirring motors. Each of the four corners rests on a rubber cushion. The powder coated steel base measures 7.875” (200mm) W x 12.5" (316mm) deep overall and has a black enamel finish. Support rod is .625” (16mm) diameter x 30” long.