Learn More
Cole-Parmer Stuart Portable Analog Stirrer; Battery Operated
Catalog No. 501960411 Shop All Cole-Parmer Products
Encompass
This compact rugged unit is ideal for use inside incubators, glove boxes or in the field. The stirrer is housed in a chemically resistant ABS plastic case and can be powered by batteries or on mains power system. Operating indicator will dim when batteries run low.
Catalog No. 50-196-0411 Supplier Cole-Parmer Supplier No. 0480105
This compact rugged unit is ideal for use inside incubators, glove boxes or in the field. The stirrer is housed in a chemically resistant ABS plastic case and can be powered by batteries or on mains power system. Operating indicator will dim when batteries run low.