Learn More
Chemglass Life Sciences Stirring Shaft, 10mm, Button Style, for 100mL to 5000mL Flasks, 455mm OAL
SDP
Catalog No. CG207501
Encompass_Preferred
Stirring Shaft, 10mm, Button Style, for 100mL to 5000mL Flasks, 455mm OAL. A 10mm precision ground stirrer shaft to fit CG-2070 through CG-2072 10mm bearings. Side button for attachment of CG-2080 or CG-2081 PTFE stirrer blades.PLEASE NOTE: THESE SHAFTS ARE NOT TO BE USED WITH ANY TYPE OF PTFE BEARING. FOR POLISHED SHAFTS USED WITH PTFE BEARINGS SEE CG-2079.
Catalog No. CG207501 Supplier Chemglass Life Sciences Supplier No. CG207501
Stirring Shaft, 10mm, Button Style, for 100mL to 5000mL Flasks, 455mm OAL. A 10mm precision ground stirrer shaft to fit CG-2070 through CG-2072 10mm bearings. Side button for attachment of CG-2080 or CG-2081 PTFE stirrer blades.PLEASE NOTE: THESE SHAFTS ARE NOT TO BE USED WITH ANY TYPE OF PTFE BEARING. FOR POLISHED SHAFTS USED WITH PTFE BEARINGS SEE CG-2079.