Learn More
Chemglass Life Sciences Stirring Shaft, 19mm, Button Style, for 1000 to 5000mL Flasks, 585mm OAL
SDP
Catalog No. 50873401
Encompass_Preferred
Stirring Shaft, 19mm, Button Style, for 1000ml. to 5000ml. Flasks, 585mm OAL. A 19mm OD precision ground stirrer shaft to fit CG-2082 and CG-2083 19mm bearings. Side button for attachment of CG-2089 PTFE stirrer blades.PLEASE NOTE: THESE SHAFTS ARE NOT TO BE USED WITH ANY TYPE OF PTFE BEARING. FOR POLISHED SHAFTS USED WITH PTFE BEARINGS SEE CG-2088. ALSO NOTE: 19MM SHAFTS HAVE A REDUCED DIAMETER TOP SECTION THAT IS 1/2"OD, PLEASE KEEP THIS IN MIND WHEN SELECTING THE PROPER SIZE COUPLING.
Catalog No. 50-873-401 Supplier Chemglass Life Sciences Supplier No. CG208501
Stirring Shaft, 19mm, Button Style, for 1000ml. to 5000ml. Flasks, 585mm OAL. A 19mm OD precision ground stirrer shaft to fit CG-2082 and CG-2083 19mm bearings. Side button for attachment of CG-2089 PTFE stirrer blades.PLEASE NOTE: THESE SHAFTS ARE NOT TO BE USED WITH ANY TYPE OF PTFE BEARING. FOR POLISHED SHAFTS USED WITH PTFE BEARINGS SEE CG-2088. ALSO NOTE: 19MM SHAFTS HAVE A REDUCED DIAMETER TOP SECTION THAT IS 1/2"OD, PLEASE KEEP THIS IN MIND WHEN SELECTING THE PROPER SIZE COUPLING.