Learn More
Chemglass Life Sciences Stirring Shaft, 10mm, Polished, Button Style, for 2000 to 12000mL Flasks, 525mm OAL
SDP
Catalog No. CG207902
Encompass_Preferred
Stirring Shaft, 10mm, Polished, Button Style, for 2000ml. to 12000ml. Flasks, 525mm OAL. A 10mm precision ground and polished stirrer shaft to fit CG-2076 and CG-2077 10mm bearings. Side button for attachment of CG-2080 or CG-2081 PTFE stirrer blades.PLEASE NOTE: THESE SHAFTS ARE NOT TO BE USED WITH ANY TYPE OF GLASS BEARING. FOR GROUND SHAFTS USED WITH GLASS BEARINGS SEE CG-2075.
Catalog No. CG207902 Supplier Chemglass Life Sciences Supplier No. CG207902
Stirring Shaft, 10mm, Polished, Button Style, for 2000ml. to 12000ml. Flasks, 525mm OAL. A 10mm precision ground and polished stirrer shaft to fit CG-2076 and CG-2077 10mm bearings. Side button for attachment of CG-2080 or CG-2081 PTFE stirrer blades.PLEASE NOTE: THESE SHAFTS ARE NOT TO BE USED WITH ANY TYPE OF GLASS BEARING. FOR GROUND SHAFTS USED WITH GLASS BEARINGS SEE CG-2075.