Learn More
Chemglass Life Sciences Stirring Shaft, 10mm, Polished, 405mm OAL
SDP
Catalog No. 50872506
Encompass_Preferred
Stirring Shaft, 10mm, Polished, 405mm OAL. A 10mm precision ground and polished stirrer shaft for use with PTFE bearings and CG-2091 and CG-2092 PTFE agitators. Flat ground section extends half way up shaft and permits positive seating of the PTFE locking screws.
Catalog No. 50-872-506 Supplier Chemglass Life Sciences Supplier No. CG2075B01
Stirring Shaft, 10mm, Polished, 405mm OAL. A 10mm precision ground and polished stirrer shaft for use with PTFE bearings and CG-2091 and CG-2092 PTFE agitators. Flat ground section extends half way up shaft and permits positive seating of the PTFE locking screws.