Learn More
Chemglass Life Sciences Stirring Shaft, 10mm, Polished, Button Style, for 5000 to 20000mL Flasks, 600mm OAL
SDP
Catalog No. CG207903
Encompass_Preferred
Stirring Shaft, 10mm, Polished, Button Style, for 5000ml. to 22000ml. Flasks, 600mm OAL. A 10mm precision ground and polished stirrer shaft to fit CG-2076 and CG-2077 10mm bearings. Side button for attachment of CG-2080 or CG-2081 PTFE stirrer blades.PLEASE NOTE: THESE SHAFTS ARE NOT TO BE USED WITH ANY TYPE OF GLASS BEARING. FOR GROUND SHAFTS USED WITH GLASS BEARINGS SEE CG-2075.
Catalog No. CG207903 Supplier Chemglass Life Sciences Supplier No. CG207903
Stirring Shaft, 10mm, Polished, Button Style, for 5000ml. to 22000ml. Flasks, 600mm OAL. A 10mm precision ground and polished stirrer shaft to fit CG-2076 and CG-2077 10mm bearings. Side button for attachment of CG-2080 or CG-2081 PTFE stirrer blades.PLEASE NOTE: THESE SHAFTS ARE NOT TO BE USED WITH ANY TYPE OF GLASS BEARING. FOR GROUND SHAFTS USED WITH GLASS BEARINGS SEE CG-2075.