Learn More
Chemglass Life Sciences Stirring Shaft, 19mm, Button Style, for 50000 to 72000mL Flasks, 950mm OAL
SDP
Catalog No. CG208503
Encompass_Preferred
Stirring Shaft, 19mm, Button Style, for 50000ml. to 72000ml. Flasks, 950mm OAL. A 19mm OD precision ground stirrer shaft to fit CG-2082 and CG-2083 19mm bearings. Side button for attachment of CG-2089 PTFE stirrer blades.PLEASE NOTE: THESE SHAFTS ARE NOT TO BE USED WITH ANY TYPE OF PTFE BEARING. FOR POLISHED SHAFTS USED WITH PTFE BEARINGS SEE CG-2088. ALSO NOTE: 19MM SHAFTS HAVE A REDUCED DIAMETER TOP SECTION THAT IS 1/2"OD, PLEASE KEEP THIS IN MIND WHEN SELECTING THE PROPER SIZE COUPLING.
Catalog No. CG208503 Supplier Chemglass Life Sciences Supplier No. CG208503
Stirring Shaft, 19mm, Button Style, for 50000ml. to 72000ml. Flasks, 950mm OAL. A 19mm OD precision ground stirrer shaft to fit CG-2082 and CG-2083 19mm bearings. Side button for attachment of CG-2089 PTFE stirrer blades.PLEASE NOTE: THESE SHAFTS ARE NOT TO BE USED WITH ANY TYPE OF PTFE BEARING. FOR POLISHED SHAFTS USED WITH PTFE BEARINGS SEE CG-2088. ALSO NOTE: 19MM SHAFTS HAVE A REDUCED DIAMETER TOP SECTION THAT IS 1/2"OD, PLEASE KEEP THIS IN MIND WHEN SELECTING THE PROPER SIZE COUPLING.