Learn More
Chemglass Life Sciences 6mm Stirrer Shaft Coupling, Universal, 1/4" OD Stud
SDP
Catalog No. CG204601
Encompass_Preferred
6mm Stirrer Shaft Coupling, Universal, 1/4" OD Stud. Coupling incorporates a universal joint to compensate for misalignment of drive motor and glass stirrer shaft. Nylon bushing compresses around shaft using an allen screw. Stainless steel stud attaches directly to the chuck of a stirrer motor. Supplied complete with an allen wrench.PLEASE NOTE: OTHER THAN OUR CG-2097 PROCESS REACTOR STIRRER SHAFTS, CHEMGLASS 19MM OD STIRRER SHAFTS HAVE A REDUCED TOP DIAMETER OF 12.7MM. WHEN ORDERING A UNIVERSAL COUPLING FOR A CHEMGLASS 19MM OD STIRRER SHAFT, USE PART NUMBER CG-2046-03.
Catalog No. CG204601 Supplier Chemglass Life Sciences Supplier No. CG204601
6mm Stirrer Shaft Coupling, Universal, 1/4" OD Stud. Coupling incorporates a universal joint to compensate for misalignment of drive motor and glass stirrer shaft. Nylon bushing compresses around shaft using an allen screw. Stainless steel stud attaches directly to the chuck of a stirrer motor. Supplied complete with an allen wrench.PLEASE NOTE: OTHER THAN OUR CG-2097 PROCESS REACTOR STIRRER SHAFTS, CHEMGLASS 19MM OD STIRRER SHAFTS HAVE A REDUCED TOP DIAMETER OF 12.7MM. WHEN ORDERING A UNIVERSAL COUPLING FOR A CHEMGLASS 19MM OD STIRRER SHAFT, USE PART NUMBER CG-2046-03.