Learn More
Chemglass Life Sciences Stirrer Shaft, Polished, 10mm, 70mm OD x 15mm Wide, 90° Four-Blade Glass Agitator, 455mm OAL
SDP
Catalog No. 5000980602
Encompass_Preferred
A 10mm precision ground stirrer shaft to fit CG-2070 through CG-2072 10mm bearings. Side button for attachment of CG-2080 or CG-2081 PTFE stirrer blades. PLEASE NOTE: THESE SHAFTS ARE NOT TO BE USED WITH ANY TYPE OF PTFE BEARING. FOR POLISHED SHAFTS USED WITH PTFE BEARINGS SEE CG-2079.
Catalog No. 50-009-80602 Supplier Chemglass Life Sciences Supplier No. CG2075U10
A 10mm precision ground stirrer shaft to fit CG-2070 through CG-2072 10mm bearings. Side button for attachment of CG-2080 or CG-2081 PTFE stirrer blades. PLEASE NOTE: THESE SHAFTS ARE NOT TO BE USED WITH ANY TYPE OF PTFE BEARING. FOR POLISHED SHAFTS USED WITH PTFE BEARINGS SEE CG-2079.