Learn More
Chemglass Life Sciences Stirrer shaft, polished glass, 19 mm, 90° four-blade agitator - 630 mm OAL (for process and filter reactors)
SDP
Catalog No. 502733531
Encompass_Preferred
Polished 19 mm diameter glass stirrer shaft with a 90° four-blade agitator, 105 mm OD and 18 mm blade width, and 630 mm overall length. Designed for process and filter reactors and to operate with PTFE bearings for reduced abrasion and vacuum-tight sealing.
  • 19 mm precision ground and polished glass shaft.
  • Four-blade 90° glass agitator with 105 mm outer diameter and 18 mm blade width.
  • Overall length 630 mm suitable for mid-size reactor vessels.
  • Designed for use with PTFE bearings to minimize abrasion and help maintain vacuum-tight seals.
  • Intended for process and filter reactors and compatible with standard reactor support frames.
Catalog No. 50-273-3531 Supplier Chemglass Life Sciences Supplier No. CG2097U50
Polished 19 mm diameter glass stirrer shaft with a 90° four-blade agitator, 105 mm OD and 18 mm blade width, and 630 mm overall length. Designed for process and filter reactors and to operate with PTFE bearings for reduced abrasion and vacuum-tight sealing.
  • 19 mm precision ground and polished glass shaft.
  • Four-blade 90° glass agitator with 105 mm outer diameter and 18 mm blade width.
  • Overall length 630 mm suitable for mid-size reactor vessels.
  • Designed for use with PTFE bearings to minimize abrasion and help maintain vacuum-tight seals.
  • Intended for process and filter reactors and compatible with standard reactor support frames.