Learn More
Chemglass Life Sciences Stirrer Shaft Coupling, 30mm, 3/8" OD Stud, Coupling Only
SDP
Supplier:  Chemglass Life Sciences CG2046X030
Encompass_Preferred
Stirrer Shaft Coupling, 30mm, 3/8" OD Stud, Coupling Only. Coupling with universal joint compensates for misalignment of drive motor and glass stir shaft.
Catalog No. 50-153-1421