Learn More
Chemglass Life Sciences Agitator, Turbine, PTFE, 100mm Dia, Fits Shaft Size 10mm
SDP
Catalog No. CG209202
Encompass_Preferred
Agitator, Turbine, PTFE, 100mm Dia, Fits Shaft Size 10mm. Agitators are made entirely of PTFE and glass filled PTFE and are for use with 10mm OD stirrer shaft CG-2075-B. Great for use a “foam breaker” at the liquid level in the reactor. Agitator hub is open so that it can be positioned anywhere along the stirrer shaft.
Catalog No. CG209202 Supplier Chemglass Life Sciences Supplier No. CG209202
Agitator, Turbine, PTFE, 100mm Dia, Fits Shaft Size 10mm. Agitators are made entirely of PTFE and glass filled PTFE and are for use with 10mm OD stirrer shaft CG-2075-B. Great for use a “foam breaker” at the liquid level in the reactor. Agitator hub is open so that it can be positioned anywhere along the stirrer shaft.