Learn More
Chemglass Life Sciences Stirrer Turbine, Upper, PTFE, Pilot Plant, 150mm OD, fits 25.4mm Shaft, 13/64in Blade Thickness, 150mm
SDP
Catalog No. CG196873
Encompass_Preferred
Stirrer Turbine, Upper, PTFE, Pilot Plant, 150mm OD, fits 25.4mm Shaft, 13/64in Blade Thickness, 150mm. PTFE six blade turbine style agitator for use with reactor stirrer shafts. Shaft opening is drilled straight through to permit positioning at any height on the shaft.
Catalog No. CG196873 Supplier Chemglass Life Sciences Supplier No. CG196873
Stirrer Turbine, Upper, PTFE, Pilot Plant, 150mm OD, fits 25.4mm Shaft, 13/64in Blade Thickness, 150mm. PTFE six blade turbine style agitator for use with reactor stirrer shafts. Shaft opening is drilled straight through to permit positioning at any height on the shaft.