Learn More
Chemglass Life Sciences Stirrer Turbine, Upper, PTFE, Pilot Plant, 100mm OD, fits 19mm Shaft, 13/64in Blade Thickness
SDP
Catalog No. CG196504
Encompass_Preferred
Stirrer Turbine, Upper, PTFE, Pilot Plant, 100mm OD, fits 19mm Shaft, 13/64in Blade Thickness. PTFE six blade turbine style agitator for use with reactor stirrer shafts. Shaft opening is drilled straight through to permit positioning at any height on the shaft.
Catalog No. CG196504 Supplier Chemglass Life Sciences Supplier No. CG196504
Stirrer Turbine, Upper, PTFE, Pilot Plant, 100mm OD, fits 19mm Shaft, 13/64in Blade Thickness. PTFE six blade turbine style agitator for use with reactor stirrer shafts. Shaft opening is drilled straight through to permit positioning at any height on the shaft.