Learn More
Chemglass Life Sciences Stir Bar, Magnetic, PTFE, Length x Dia. in mm: 22.2 x 9.52, Length x Dia. in inches: 7/8 x 3/8
SDP
Catalog No. 501531389
Encompass_Preferred
Stir Bar, Magnetic, PTFE, Length x Dia. in mm: 22.2 x 9.52, Length x Dia. in inches: 7/8 x 3/8. Octagon cross-section having a moulded-in pivot ring manufactured with FDA-grade and USP Class VI PTFE. The PTFE coated Alnico magnet permits magnetic agitation without contamination.
Catalog No. 50-153-1389 Supplier Chemglass Life Sciences Supplier No. CG200107
Stir Bar, Magnetic, PTFE, Length x Dia. in mm: 22.2 x 9.52, Length x Dia. in inches: 7/8 x 3/8. Octagon cross-section having a moulded-in pivot ring manufactured with FDA-grade and USP Class VI PTFE. The PTFE coated Alnico magnet permits magnetic agitation without contamination.