Learn More
Chemglass Life Sciences Chemglass Stirring Bar, Plain PTFE Magnetic, 30 x 6 mm, for Magnetic Stirrers
SDP
Catalog No. NC2748315
Encompass_Preferred
The Chemglass Stirring Bar (plain) is a PTFE-coated magnetic stir bar sized 30 x 6 mm, designed for reliable mixing in laboratory vessels. Its plain profile creates more turbulence compared with cylindrical bars at low speeds, improving mixing efficiency on magnetic stir plates.
  • PTFE construction provides chemical resistance and temperature stability.
  • Plain shape increases turbulence at low stirring speeds.
  • Dimensions: 30 x 6 mm, suitable for small-to-medium vessels.
  • Magnetic core for use with magnetic stir plates.
  • Sold individually (each).
Catalog No. NC2748315 Supplier Chemglass Life Sciences Supplier No. CG2001P05
The Chemglass Stirring Bar (plain) is a PTFE-coated magnetic stir bar sized 30 x 6 mm, designed for reliable mixing in laboratory vessels. Its plain profile creates more turbulence compared with cylindrical bars at low speeds, improving mixing efficiency on magnetic stir plates.
  • PTFE construction provides chemical resistance and temperature stability.
  • Plain shape increases turbulence at low stirring speeds.
  • Dimensions: 30 x 6 mm, suitable for small-to-medium vessels.
  • Magnetic core for use with magnetic stir plates.
  • Sold individually (each).