Learn More
Chemglass Life Sciences Stir Bar Dual Purpose Positioner/Retriever, Polyethylene, 5/8" Diameter X 12" OAL
SDP
Catalog No. CG201901
Encompass_Preferred
Stir Bar Dual Purpose Positioner/Retriever, Polyethylene, 5/8" Diameter X 12" OAL. Convenient dual purpose polyethylene unit permits positioning of the spinbar at the bottom of a vessel without dropping bar into liquid. Also allows bar to be removed without touching or pouring out liquid. Retriever slides within outer casing. To pick up spinbar, slide retriever down; to release bar, slide retriever up. 5/8in in diameter and 12in overall length.
Catalog No. CG201901 Supplier Chemglass Life Sciences Supplier No. CG201901
Stir Bar Dual Purpose Positioner/Retriever, Polyethylene, 5/8" Diameter X 12" OAL. Convenient dual purpose polyethylene unit permits positioning of the spinbar at the bottom of a vessel without dropping bar into liquid. Also allows bar to be removed without touching or pouring out liquid. Retriever slides within outer casing. To pick up spinbar, slide retriever down; to release bar, slide retriever up. 5/8in in diameter and 12in overall length.