Learn More
Cole-Parmer Starburst Magnetic Stirring Heads, 3/4" x 3/4"; 1/Pk
Supplier:  Cole-Parmer 0476512
Encompass_Preferred
Stirring heads produce a large vortex for quick, efficient mixing—conserve both energy and valuable lab time. Powerful alnico V magnet is coated with PTFE for purity and excellent chemical resistance. Fully autoclavable.
Catalog No. NC0585075