Learn More
Med Vet International Stirring Hot Plate
Catalog No. 503047038
Encompass_Preferred
With this stirrer hot plate you can heat and stir simultaneously or you can perform each separately. Stirrer hot plate can carry out precise stable stirring of the liquid even at high speeds. Comes with Tempature probe. LED display can manually switch between set tempature and actual temperature.
Catalog No. 50-304-7038 Supplier Med Vet International Supplier No. 881
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
With this stirrer hot plate you can heat and stir simultaneously or you can perform each separately. Stirrer hot plate can carry out precise stable stirring of the liquid even at high speeds. Comes with Tempature probe. LED display can manually switch between set tempature and actual temperature.