Learn More
Electron Microscopy Sciences Magnetic Stirring and Mixing Bars, 41.3 mm L, 8 mm diameter
SDP
Catalog No. 5029906
Encompass_Preferred
These bars are designed for single use applications where efficiency and cross contamination is a concern. These economical bars have a "PTFE" coating which is chemically inert for high purity contact. The magnet is an Alnico V.
OAL: 41.3 mm
Diameter: 8 mm
Catalog No. 50-299-06 Supplier Electron Microscopy Sciences Supplier No. 6452116
These bars are designed for single use applications where efficiency and cross contamination is a concern. These economical bars have a "PTFE" coating which is chemically inert for high purity contact. The magnet is an Alnico V.
OAL: 41.3 mm
Diameter: 8 mm