Learn More
Electron Microscopy Sciences Stirring Rod PTFE with Steel Core, 400 mm, 6 mm diameter
SDP
Catalog No. 5032646
Encompass_Preferred
Steel core can be bent into a permanent shape, rods are inert and will not scratch, use up to 280 degree C, rod OD: 6 mm.
OAL: 400 mm
Rod: 6 mm
Catalog No. 50-326-46 Supplier Electron Microscopy Sciences Supplier No. 6620040
Steel core can be bent into a permanent shape, rods are inert and will not scratch, use up to 280 degree C, rod OD: 6 mm.
OAL: 400 mm
Rod: 6 mm