Learn More
Electron Microscopy Sciences Stirring Rod Solid PTFE Rod, 100 mm, 8 mm diameter
SDP
Catalog No. 5032645
Encompass_Preferred
PTFE solid rod with tapered end, rods are inert and will not scratch. Use up to 280 deg C.
OAL: 100 mm
Rod: 8 mm
Catalog No. 50-326-45 Supplier Electron Microscopy Sciences Supplier No. 6620110
PTFE solid rod with tapered end, rods are inert and will not scratch. Use up to 280 deg C.
OAL: 100 mm
Rod: 8 mm