Learn More
Electron Microscopy Sciences Stirring Rod Solid PTFE Rod, 250 mm, 8 mm diameter
SDP
Catalog No. 5032647
Encompass_Preferred
PTFE solid rod with tapered end, rods are inert and will not scratch, use up to 280 deg C.
OAL: 250 mm
Rod: 8 mm
Catalog No. 50-326-47 Supplier Electron Microscopy Sciences Supplier No. 6620125
PTFE solid rod with tapered end, rods are inert and will not scratch, use up to 280 deg C.
OAL: 250 mm
Rod: 8 mm