Learn More
Chemglass Life Sciences Stirring Shaft, 10mm, Chem-Stir, 100mL-5L Flasks, 468mm OAL, Complete with PTFE Nut and Bolt
SDP
Catalog No. CG207401
Encompass_Preferred
Stirring Shaft, 10mm, Chem-Stir, 100mL-5L Flasks, 468mm OAL, Complete with PTFE Nut and Bolt.¿ PTFE is the only contact in the lower radius of the flask, therefore breakage due to dropping the stirrer shaft has been eliminated.¿ Design permits the proper amount of tension to be applied to the blade, thereby preventing the blade from flipping over to the incorrect side during flask insertion.¿ Design permits use with any PTFE stirrer blade with the keyhole shaped cutout for 10mm or 19mm shafts.A 10mm OD stirrer shaft with our patented* Chem-Stir blade attachment system. Shaft has been manufactured w/ ring at the lower end to permit attachment of the blade with a PTFE nut and bolt. Shaft is supplied complete with PTFE nut and bolt. PLEASE NOTE: STIRRER BLADES MUST BE PURCHASED SEPARATELY.THESE SHAFTS ARE NOT TO BE USED WITH ANY TYPE OF PTFE BEARING. FOR POLISHED SHAFTS USED WITH PTFE BEARINGS SEE CG-2078.* US Patent Number 5,326,167
Catalog No. CG207401 Supplier Chemglass Life Sciences Supplier No. CG207401
Stirring Shaft, 10mm, Chem-Stir, 100mL-5L Flasks, 468mm OAL, Complete with PTFE Nut and Bolt.¿ PTFE is the only contact in the lower radius of the flask, therefore breakage due to dropping the stirrer shaft has been eliminated.¿ Design permits the proper amount of tension to be applied to the blade, thereby preventing the blade from flipping over to the incorrect side during flask insertion.¿ Design permits use with any PTFE stirrer blade with the keyhole shaped cutout for 10mm or 19mm shafts.A 10mm OD stirrer shaft with our patented* Chem-Stir blade attachment system. Shaft has been manufactured w/ ring at the lower end to permit attachment of the blade with a PTFE nut and bolt. Shaft is supplied complete with PTFE nut and bolt. PLEASE NOTE: STIRRER BLADES MUST BE PURCHASED SEPARATELY.THESE SHAFTS ARE NOT TO BE USED WITH ANY TYPE OF PTFE BEARING. FOR POLISHED SHAFTS USED WITH PTFE BEARINGS SEE CG-2078.* US Patent Number 5,326,167