Learn More
Chemglass Life Sciences Stirring Shaft, 10mm, Polished, Glass Agitator, Three-Blade, 400mm OAL, for 100mL to 250mL Reactors, 42mm OD Agitator
SDP
Catalog No. CG2079A01
Encompass_Preferred
Stirring Shaft, 10mm, Polished, Glass Agitator, Three-Blade, 400mm OAL, for 100mL to 250mL Reactors, 42mm OD Agitator. Shaft is precision ground and polished to fit our CG-2076 and CG-2077 10mm PTFE bearings. Designed to be used with cylindrical type reactors. The curved glass blades are spaced 120° apart and angled upward. PLEASE NOTE: STIRRER SHAFTS ARE DESIGNED FOR COUNTER-CLOCKWISE ROTATION.
Catalog No. CG2079A01 Supplier Chemglass Life Sciences Supplier No. CG2079A01
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Stirring Shaft, 10mm, Polished, Glass Agitator, Three-Blade, 400mm OAL, for 100mL to 250mL Reactors, 42mm OD Agitator. Shaft is precision ground and polished to fit our CG-2076 and CG-2077 10mm PTFE bearings. Designed to be used with cylindrical type reactors. The curved glass blades are spaced 120° apart and angled upward. PLEASE NOTE: STIRRER SHAFTS ARE DESIGNED FOR COUNTER-CLOCKWISE ROTATION.