Learn More
Chemglass Life Sciences 10mm Stirring Shaft, Chem-Stir, Polished, For Use With 5L-20L Flasks, 605mm OAL, Complete with PTFE Nut and Bolt
SDP
Catalog No. 50872511
Encompass_Preferred
10mm Stirring Shaft, Chem-Stir, Polished, For Use With 5L-22L Flasks, 605mm OAL, Complete with PTFE Nut and Bolt. A 10mm OD precision ground and polished stirrer shaft with our patented** Chem-Stir blade attachment system. Shaft (C) has been manufactured with a ring at the lower end to permit attachment of the blade (B) with a PTFE nut (D) and bolt (A). Shaft is supplied complete with PTFE nut and bolt set. For use with PTFE bearings such as the CG-2077. Polished shaft will not cause abrasion to the sealing surfaces of the PTFE bearing, thereby allowing a vacuum tight seal. Uses the same PTFE nut and bolt blade attachment method as the CG-2074 ground shaft. PLEASE NOTE: STIRRER BLADES MUST BE PURCHASED SEPARATELY.THESE SHAFTS ARE NOT TO BE USED WITH ANY TYPE OF GLASS BEARING. FOR GROUND SHAFTS USED WITH GLASS BEARINGS SEE CG-2074.
Catalog No. 50-872-511 Supplier Chemglass Life Sciences Supplier No. CG207803
10mm Stirring Shaft, Chem-Stir, Polished, For Use With 5L-22L Flasks, 605mm OAL, Complete with PTFE Nut and Bolt. A 10mm OD precision ground and polished stirrer shaft with our patented** Chem-Stir blade attachment system. Shaft (C) has been manufactured with a ring at the lower end to permit attachment of the blade (B) with a PTFE nut (D) and bolt (A). Shaft is supplied complete with PTFE nut and bolt set. For use with PTFE bearings such as the CG-2077. Polished shaft will not cause abrasion to the sealing surfaces of the PTFE bearing, thereby allowing a vacuum tight seal. Uses the same PTFE nut and bolt blade attachment method as the CG-2074 ground shaft. PLEASE NOTE: STIRRER BLADES MUST BE PURCHASED SEPARATELY.THESE SHAFTS ARE NOT TO BE USED WITH ANY TYPE OF GLASS BEARING. FOR GROUND SHAFTS USED WITH GLASS BEARINGS SEE CG-2074.