Learn More
Chemglass Life Sciences Stirring Shaft, 10mm, Polished, Glass Agitator, Three-Blade, 445mm OAL, for 500mL Reactors, 66mm OD Agitator
SDP
Catalog No. 50997816
Encompass_Preferred
Stirring Shaft, 10mm, Polished, Glass Agitator, Three-Blade, 445mm OAL, for 500mL Reactors, 66mm OD Agitator. Shaft is precision ground and polished to fit our CG-2076 and CG-2077 10mm PTFE bearings. Designed to be used with cylindrical type reactors. The curved glass blades are spaced 120° apart and angled upward. PLEASE NOTE: STIRRER SHAFTS ARE DESIGNED FOR COUNTER-CLOCKWISE ROTATION.
Catalog No. 50-997-816 Supplier Chemglass Life Sciences Supplier No. CG2079A02
Stirring Shaft, 10mm, Polished, Glass Agitator, Three-Blade, 445mm OAL, for 500mL Reactors, 66mm OD Agitator. Shaft is precision ground and polished to fit our CG-2076 and CG-2077 10mm PTFE bearings. Designed to be used with cylindrical type reactors. The curved glass blades are spaced 120° apart and angled upward. PLEASE NOTE: STIRRER SHAFTS ARE DESIGNED FOR COUNTER-CLOCKWISE ROTATION.