Learn More
Chemglass Life Sciences Stirring Shaft, PTFE Covered SS, 458mm OAL, 66mm OD Retreating Blade, Fits Reactors up to 500mL
SDP
Catalog No. 501531441
Encompass_Preferred
Stirring Shaft, PTFE Covered SS, 458mm OAL, 66mm OD Retreating Blade, Fits Reactors up to 500mL. 10mm O.D. shaft is constructed using a molding process whereas the blade is molded directly onto the PTFE coated Stainless Steel shaft. This makes the stirrer chemically inert, easy to clean and can be used up to 280C. The top of the shaft has 50mm of exposed 1/4" diameter Stainless Steel for rigid clamping to a coupling such as CG-2044-C-10 or CG-2046-01.Note: Shaft must be used with CG-2077-U or CG-2077-M style bearing
Catalog No. 50-153-1441 Supplier Chemglass Life Sciences Supplier No. CG2079X050
Stirring Shaft, PTFE Covered SS, 458mm OAL, 66mm OD Retreating Blade, Fits Reactors up to 500mL. 10mm O.D. shaft is constructed using a molding process whereas the blade is molded directly onto the PTFE coated Stainless Steel shaft. This makes the stirrer chemically inert, easy to clean and can be used up to 280C. The top of the shaft has 50mm of exposed 1/4" diameter Stainless Steel for rigid clamping to a coupling such as CG-2044-C-10 or CG-2046-01.Note: Shaft must be used with CG-2077-U or CG-2077-M style bearing