Learn More
Aviva Systems Biology Corp STK11 Antibody - N-terminal region (ARP41743_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41743_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against STK11. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human STK11. Alternate names: LKB1, PJS, hLKB1ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-57977