Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC STK11 Human His-SU 10ug
SDP

Catalog No. 5000195887
Encompass_Preferred

Recombinant human STK11 (LKB1) is a serine/threonine kinase that regulates AMPK family members and modulates cellular metabolism, apoptosis, and DNA damage responses. Supplied lyophilized and expressed in E. coli with N-terminal SUMO and 6xHis tags, it is intended for biochemical characterization, antibody validation, and functional studies.

  • Recombinant human kinase for biochemical assays.
  • Expressed in E. coli for high yield production.
  • N-terminal SUMO and 6xHis tags for purification and detection.
  • Approximate molecular weight 60-72 kDa.
  • Supplied lyophilized with stabilizing trehalose formulation.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Stable long-term at -20°C; aliquot and freeze for extended storage.

Catalog No. 50-001-95887 Supplier Medchemexpress LLC Supplier No. HYP7154910UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human STK11 (LKB1) is a serine/threonine kinase that regulates AMPK family members and modulates cellular metabolism, apoptosis, and DNA damage responses. Supplied lyophilized and expressed in E. coli with N-terminal SUMO and 6xHis tags, it is intended for biochemical characterization, antibody validation, and functional studies.

  • Recombinant human kinase for biochemical assays.
  • Expressed in E. coli for high yield production.
  • N-terminal SUMO and 6xHis tags for purification and detection.
  • Approximate molecular weight 60-72 kDa.
  • Supplied lyophilized with stabilizing trehalose formulation.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Stable long-term at -20°C; aliquot and freeze for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.