Learn More
Medchemexpress LLC STK11 Human His-SU 50ug
SDP
Catalog No. 5000195888
Encompass_Preferred
STK11 Protein, Human (His-SUMO) is a recombinant human serine/threonine kinase (LKB1) expressed in E. coli and supplied lyophilized for research applications. The protein includes N-terminal SUMO and 6xHis tags to facilitate purification and detection, covers residues M1-C430, and is purified to greater than 90% as determined by reducing SDS-PAGE. It is provided in a stabilizing formulation and should be stored frozen for long-term stability.
  • Recombinant human serine/threonine kinase covering residues M1-C430.
  • N-terminal SUMO and 6xHis tags for purification and detection.
  • Expressed in E. coli and purified to greater than 90% purity by reducing SDS-PAGE.
  • Supplied lyophilized in a stabilizing buffer for storage and transport.
  • Stable when stored at -20°C to -80°C; reconstituted material is stable short term at 4°C.
  • Intended for research use only; not for human or diagnostic use.
Catalog No. 50-001-95888 Supplier Medchemexpress LLC Supplier No. HYP7154950UG
STK11 Protein, Human (His-SUMO) is a recombinant human serine/threonine kinase (LKB1) expressed in E. coli and supplied lyophilized for research applications. The protein includes N-terminal SUMO and 6xHis tags to facilitate purification and detection, covers residues M1-C430, and is purified to greater than 90% as determined by reducing SDS-PAGE. It is provided in a stabilizing formulation and should be stored frozen for long-term stability.
  • Recombinant human serine/threonine kinase covering residues M1-C430.
  • N-terminal SUMO and 6xHis tags for purification and detection.
  • Expressed in E. coli and purified to greater than 90% purity by reducing SDS-PAGE.
  • Supplied lyophilized in a stabilizing buffer for storage and transport.
  • Stable when stored at -20°C to -80°C; reconstituted material is stable short term at 4°C.
  • Intended for research use only; not for human or diagnostic use.